Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl Red Indicator, 0.1% w/v in Ethanol, Reagents
Click to view available options
Quantity:
120 mL
500 mL
Specifications
Specifications
Chemical Name or Material | Methyl Red Indicator, 0.1% w/v in Ethanol |
CAS | 64-17-5, 7732-18-5, 7732-18-5 |
Boiling Point | 77°C |
Synonym | Acid Red 2, 2-[[4-(Dimethylamino)phenyl]diazenyl]benzoic Acid |
Percent Purity | 0.10% |
Grade | Reagent |
Packaging | Poly Bottle |
Specific Gravity | 0.8 |
Color | Red |
Quantity | 120 mL |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction