Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Red, Sodium Salt, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
10g
25g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

845-10-3
C15H14N3NaO2
291.286
GNTPCYMJCJNRQB-UHFFFAOYSA-M
C.I. 13020
4465632
sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 291.29
Packaging Wide Mouth Amber Glass Bottle
Quantity 10g
Chemical Name or Material Methyl Red, Sodium Salt
Grade Baker Analyzed™ ACS Reagent