Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (S)-3-hydroxy-3-phenylpropanoate, 99%, Thermo Scientific™

Catalog Number ac414450010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
36615-45-9
C10H12O3
180.2
MFCD00221509
VZHHCQFCDCJKIX-VIFPVBQESA-N
s-methyl 3-hydroxy-3-phenylpropanoate, methyl s-3-hydroxy-3-phenylpropanoate, methyl 3s-3-hydroxy-3-phenylpropanoate, methyl s-3-hydroxy-3-phenylpropionate, methyl s---3-hydroxy-3-phenylpropionate, benzenepropanoic acid, b-hydroxy-, methylester, bs
6993014
methyl (3S)-3-hydroxy-3-phenylpropanoate
COC(=O)CC(C1=CC=CC=C1)O
36615-45-9
C10H12O3
180.2
MFCD00221509
VZHHCQFCDCJKIX-VIFPVBQESA-N
s-methyl 3-hydroxy-3-phenylpropanoate, methyl s-3-hydroxy-3-phenylpropanoate, methyl 3s-3-hydroxy-3-phenylpropanoate, methyl s-3-hydroxy-3-phenylpropionate, methyl s---3-hydroxy-3-phenylpropionate, benzenepropanoic acid, b-hydroxy-, methylester, bs
6993014
methyl (3S)-3-hydroxy-3-phenylpropanoate
COC(=O)CC(C1=CC=CC=C1)O
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5180 to 1.52
Quantity 1 g
Specific Rotation Condition − 44.20 (21.00°C c=3,1,2−dichloroeth)
Specific Rotation -44.2
Formula Weight 180.2
Percent Purity 99%
Physical Form Viscous Liquid
Chemical Name or Material Methyl (S)-3-hydroxy-3-phenylpropanoate, 99%
Show More Show Less

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit