Alfa Aesar™ Methyl salicylate, 98%

Manufacturer: Alfa Aesar™ A1057836

Quantity 500g

 View more versions of this product

Catalog No. AAA1057836

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 119-36-8
    Molecular Formula C8H8O3
    Formula Weight 152.15
    MDL Number MFCD00002214
    Beilstein 971516
    Merck Index 14,6120
    Synonym Methyl 2-hydroxybenzoate; Salicylic acid methyl ester
    Chemical Name or Material Methyl salicylate

    Specifications

    Formula Weight 152.15
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 119-36-8
    MDL Number MFCD00002214
    Beilstein 971516
    TSCA TSCA
    Odor Characteristic
    Merck Index 14,6120
    Melting Point −9°C
    Boiling Point 224°C
    Flash Point 99°C (210°F)
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number VO4725000
    Quantity 500g
    Molecular Formula C8H8O3
    Synonym Methyl 2-hydroxybenzoate; Salicylic acid methyl ester
    Health Hazard 1 Warning
    Chemical Name or Material Methyl salicylate
    Assay 98%

    Description

    Applications
    Methyl salicylate is used as a flavoring agent and providing fragrance to products like chewing gum and mints. It acts as an anticoagulant by interruption of the vitamin K1-epoxide cycle. It is also used in high concentrations as a rubefacient to treat joint, muscular pain and acute pain. It is utilized as an anti-herbivore defense system in various plants. It is used as a transfer agent to get a manual copy of an image on a surface and to clear plant or animal tissue samples to be used for microscopy.

    Solubility
    Miscible with water, chloroform, ethanol, methonol and ether.

    Notes
    Light sensitive. Incompatible with strong oxidizing agents and strong bases.

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit