Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Sulfide, +99%, Extra Pure, ACROS Organics™

Click to view available options
Quantity:
10mL
1L
2.5L
Packaging:
Glass bottle
75-18-3
C2H6S
62.13
MFCD00008562
QMMFVYPAHWMCMS-UHFFFAOYSA-N
dimethyl sulfide,methyl sulfide,methane, thiobis,dimethylsulfide,dimethylsulphide,methyl thioether,2-thiapropane,dimethyl sulphide,dimethyl thioether,methylthiomethane
1068
CHEBI:17437
methylsulfanylmethane
CSC
75-18-3
C2H6S
62.13
MFCD00008562
QMMFVYPAHWMCMS-UHFFFAOYSA-N
dimethyl sulfide,methyl sulfide,methane, thiobis,dimethylsulfide,dimethylsulphide,methyl thioether,2-thiapropane,dimethyl sulphide,dimethyl thioether,methylthiomethane
1068
CHEBI:17437
methylsulfanylmethane
CSC
Density 0.8460g/mL
Refractive Index 1.4351
Water 0.3% max.
Quantity 10mL
Melting Point -98.0°C
Infrared Spectrum Authentic
Boiling Point 38.0°C
Color Colorless
Flash Point −48°C
Packaging Glass bottle
Specific Gravity 0.846
Assay Percent Range 99+%
Linear Formula (CH3)2S
Beilstein 01, 288
Fieser 02,156; 04,190; 05,260; 15,88; 16,84; 17,198
Merck Index 15, 3281
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 62.13
Physical Form Liquid
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material Methyl sulfide
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger