Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl sulfone, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
500g
2.5kg
Packaging:
Glass Bottle
Plastic bottle
67-71-0
C2H6O2S
94.128
HHVIBTZHLRERCL-UHFFFAOYSA-N
dimethyl sulfone,methyl sulfone,sulfonylbismethane,dimethylsulfone,dimethyl sulphone,methane, sulfonylbis,sulphonylbismethane,methylsulfone,methanesulfonylmethane,sulfonyldimethane
6213
CHEBI:9349
methylsulfonylmethane
CS(=O)(=O)C
67-71-0
C2H6O2S
94.128
HHVIBTZHLRERCL-UHFFFAOYSA-N
dimethyl sulfone,methyl sulfone,sulfonylbismethane,dimethylsulfone,dimethyl sulphone,methane, sulfonylbis,sulphonylbismethane,methylsulfone,methanesulfonylmethane,sulfonyldimethane
6213
CHEBI:9349
methylsulfonylmethane
CS(=O)(=O)C
Color White
Infrared Spectrum Authentic
Melting Point 107.0°C to 111.0°C
Boiling Point 238.0°C
Packaging Plastic bottle
Quantity 2.5kg
Flash Point 143°C
Assay Percent Range 98%
Linear Formula (CH3)2SO2
Formula Weight 94.13
Physical Form Crystals or Crystalline Powder
Percent Purity 98%
Chemical Name or Material Methyl sulfone
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.