Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl sulfoxide, 99.7%, Extra Dry, anhydrous, SC, AcroSeal™, ACROS Organics™ Edge Available on GSA/VA Contract for Federal Government customers only.
Edge

Click to view available options
Quantity:
100mL
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
methanesulfinylmethane
CS(C)=O
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
methanesulfinylmethane
CS(C)=O

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Additional Information Vapor Pressure: 0.55mbar at 20°C
Assay Percent Range 99.7%
Linear Formula (CH3)2SO
Beilstein 01, IV, 1277
Fieser 01,296; 02,157; 03,119; 04,192; 05,263; 06,225; 07,133; 08,198; 09,189; 10,166; 12,212; 13,124; 15,146; 16,149; 17,127
Merck Index 15, 3285
Formula Weight 78.13
Percent Purity ≥99.7%
Grade Extra Dry
Density 2.7 (Air = 1.0)
Identification Pass Test
Water 50ppm max.
Decomposition Information >190°C
Quantity 100mL
Chemical Name or Material Methyl sulfoxide
Show More Show Less