Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl sulfoxide, 99.7+%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
1L
2.5L
Packaging:
AcroSeal glass Bottle
AcroSeal™ Glass Bottle
Glass bottle
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
methylsulfinylmethane
CS(C)=O
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
methylsulfinylmethane
CS(C)=O
Water 0.005% Max.
Vapor Pressure 0.55mbar at 20°C
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4770 to 1.4800
Specific Gravity 1.1
Quantity 100mL
Infrared Spectrum Authentic
Decomposition Information >190°C
Flash Point 87°C
Melting Point 18.4°C
Boiling Point 189.0°C
Density 1.1000g/mL
Color Brown to White
Assay Percent Range 99.7+%
Linear Formula (CH3)2SO
Beilstein 01, IV, 1277
Fieser 01,296; 02,157; 03,119; 04,192; 05,263; 06,225; 07,133; 08,198; 09,189; 10,166; 12,212; 13,124; 15,146; 16,149; 17,127
Merck Index 15, 3285
Formula Weight 78.13
Physical Form Powder, Needles or Crystals
Percent Purity ≥99.7%
Grade Extra Dry over Molecular Sieve
Chemical Name or Material Methyl sulfoxide
Show More Show Less