Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl sulfoxide, 99.8+%, for peptide synthesis, ACROS Organics™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
Glass bottle
67-68-5
C2H6OS
78.129
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide,dmso,methyl sulfoxide,dimethylsulfoxide,dimethyl sulphoxide,methane, sulfinylbis,demsodrox,demasorb,demavet,dimexide
679
CHEBI:28262
methylsulfinylmethane
CS(=O)C
67-68-5
C2H6OS
78.129
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide,dmso,methyl sulfoxide,dimethylsulfoxide,dimethyl sulphoxide,methane, sulfinylbis,demsodrox,demasorb,demavet,dimexide
679
CHEBI:28262
methylsulfinylmethane
CS(=O)C
Acetic Acid 0.003% max.
Assay Percent Range 99.8+%
Linear Formula (CH3)2SO
Beilstein 01, IV, 1277
Fieser 01,296; 02,157; 03,119; 04,192; 05,263; 06,225; 07,133; 08,198; 09,189; 10,166; 12,212; 13,124; 15,146; 16,149; 17,127
Merck Index 15, 3285
Formula Weight 78.13
Physical Form Liquid at 20°C
Percent Purity ≥99.8;%
Density 1.1000g/mL
Free Amine 0.0002% max.
Packaging Glass bottle
Refractive Index 1.4770 to 1.4800
Residue after Evaporation 0.001% max.
Solubility Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene, acetone, chloroform,, diethyl ether and aromatic hydrocarbons
Specific Gravity 1.1
Water 0.005% max. (K.F.)
Flash Point 87°C
Viscosity 1.987 mPa.s (25°C)
Boiling Point 189.0°C
Color Colorless
Melting Point 18.4°C
Quantity 2.5L
Chemical Name or Material Methyl sulfoxide, For peptide synthesis
Show More Show Less