Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl tert-Butyl Ether, HPLC Grade, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

1634-04-4
C5H12O
88.15
BZLVMXJERCGZMT-UHFFFAOYSA-N
15413
CHEBI:27642

High purity solvents for use in analytical and preparative separations

    For use in liquid chromatography (HPLC & UHPLC) & spectrophotometry
Specific Gravity 1L = 0.74kg
Quantity 1L
Flash Point −27.00°C
CAS 1634-04-4
Packaging Narrow Mouth Amber Glass Bottle
Molecular Formula C5H12O
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
SMILES CC(C)(C)OC
IUPAC Name 2-methoxy-2-methylpropane
Molecular Weight (g/mol) 88.15
PubChem CID 15413
ChEBI CHEBI:27642
Formula Weight 88.15
Chemical Name or Material Methyl tert-Butyl Ether
Grade HPLC Grade
Show More Show Less