Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl tert-Butyl Ether, ULTRA RESI-ANALYZED™, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

1634-04-4
C5H12O
88.15
BZLVMXJERCGZMT-UHFFFAOYSA-N
15413
CHEBI:27642
2-methoxy-2-methylpropane
CC(C)(C)OC

High purity solvents and salts developed for organic residue extraction/concentration procedures

    For use in Gas Chromatography (GC), Organic Residue Analysis; For Laboratory, Research, or Manufacturing Use
Specific Gravity 1L = 0.74kg
Flash Point −27.00°C
Quantity 4L
Packaging Narrow Mouth Amber Glass Bottle
Formula Weight 88.15
Chemical Name or Material Methyl tert-Butyl Ether
Grade ULTRA RESI-ANALYZED™