Alfa Aesar™ Methyl tetradecanoate, 99%

Manufacturer: Alfa Aesar™ A1025718

Quantity 50g

 View more versions of this product

Catalog No. AAA1025718

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 124-10-7
    Molecular Formula C15H30O2
    Linear Formula CH3(CH2)12CO2CH3
    Formula Weight 242.4
    MDL Number MFCD00008983
    Synonym Methyl myristate; Tetradecanoic acid methyl ester
    Chemical Name or Material Methyl tetradecanoate

    Specifications

    Formula Weight 242.4
    Boiling Point 323°C
    Molecular Formula C15H30O2
    Linear Formula CH3(CH2)12CO2CH3
    MDL Number MFCD00008983
    CAS 124-10-7
    Flash Point >112°C (234°F)
    Melting Point 18°C
    Quantity 50g
    TSCA TSCA
    Synonym Methyl myristate; Tetradecanoic acid methyl ester
    Chemical Name or Material Methyl tetradecanoate
    Assay 99%

    Description

    Applications
    Methyl tetradecanoate is used as a component in the preparation of a high-density biodiesel. Also used as a niosome medium in which an anti-cancer drug could be transported within.

    Solubility
    Soluble in chloroform and methanol. React with water.

    Notes
    Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit