Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl thiazole-2-carboxylate, 95%

Click to view available options
Quantity:
250mg
1g
5g
55842-56-3
C5H5NO2S
143.16
MFCD18075580
YRMZUOPMHMPNPG-UHFFFAOYSA-N
methyl thiazole-2-carboxylate,methyl 2-thiazolecarboxylate,2-thiazolecarboxylic acid methyl ester,methyl2-thiazolecarboxylate
7567660
methyl 1,3-thiazole-2-carboxylate
COC(=O)C1=NC=CS1
55842-56-3
C5H5NO2S
143.16
MFCD18075580
YRMZUOPMHMPNPG-UHFFFAOYSA-N
methyl thiazole-2-carboxylate,methyl 2-thiazolecarboxylate,2-thiazolecarboxylic acid methyl ester,methyl2-thiazolecarboxylate
7567660
methyl 1,3-thiazole-2-carboxylate
COC(=O)C1=NC=CS1
Quantity 1g
Formula Weight 143.16
Physical Form Solid
Percent Purity 95%
Chemical Name or Material Methyl thiazole-2-carboxylate

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures