Alfa Aesar™ Methyl tiglate, 98%

Manufacturer: Alfa Aesar™ A1196409

Quantity 10g

 View more versions of this product

Catalog No. AAA1196409

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6622-76-0
    Molecular Formula C6H10O2
    Formula Weight 114.15
    MDL Number MFCD00016654
    UN Number UN3272
    Beilstein 1720455
    Merck Index 14,9433
    Synonym Methyl trans-2,3-dimethylacrylate; Methyl (E)-2-methyl-2-butenoic acid
    Chemical Name or Material Methyl tiglate

    Specifications

    Formula Weight 114.15
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C6H10O2
    CAS 6622-76-0
    UN Number UN3272
    Flash Point 39°C (102°F)
    Beilstein 1720455
    Synonym Methyl trans-2,3-dimethylacrylate; Methyl (E)-2-methyl-2-butenoic acid
    Health Hazard 1 Warning
    Boiling Point 138° to 139°C
    Quantity 10g
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapor.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
    RTECS Number EM9254600
    TSCA TSCA
    MDL Number MFCD00016654
    Merck Index 14,9433
    Chemical Name or Material Methyl tiglate
    Assay 98%

    Description

    Applications
    It is employed as a perfuming agent in cosmetics industry.

    Solubility
    Soluble in alcohol, water 3813 mg/L @ 25°C (est).

    Notes
    Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.

    GHS H Statement
    H226
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit