Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl trans-3-(benzyloxycarbonylamino)cyclohexanecarboxylate, 97%, Thermo Scientific™

Catalog No. AAH6256003
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1035325-22-4
Molecular Formula C16H21NO4
Molecular Weight (g/mol) 291.347
MDL Number MFCD21365071
InChI Key OGLDGYYJEHFVKN-KGLIPLIRSA-N
Synonym trans-methyl-3-benzyloxy carbonyl-amino cyclohexanecarboxylate
PubChem CID 118309547
IUPAC Name methyl (1R,3S)-3-(phenylmethoxycarbonylamino)cyclohexane-1-carboxylate
SMILES COC(=O)C1CCCC(C1)NC(=O)OCC2=CC=CC=C2

Specifications

Quantity 1 g
Formula Weight 291.34
Percent Purity 97%
Chemical Name or Material Methyl trans-3-(benzyloxycarbonylamino)cyclohexanecarboxylate
TSCA No
Recommended Storage Ambient temperatures
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.