Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (trimethylsilyl)acetate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
2916-76-9
C6H14O2Si
146.261
MFCD00008450
JIHUZDFFYVRXKP-UHFFFAOYSA-N
methyl trimethylsilyl acetate,methyl trimethylsilylacetate,methyl 2-trimethylsilyl acetate,acetic acid, trimethylsilyl-, methyl ester,acmc-1ch43,jihuzdffyvrxkp-uhfffaoysa,trimethylsilanyl-acetic acid methyl ester
76210
methyl 2-trimethylsilylacetate
COC(=O)C[Si](C)(C)C
2916-76-9
C6H14O2Si
146.261
MFCD00008450
JIHUZDFFYVRXKP-UHFFFAOYSA-N
methyl trimethylsilyl acetate,methyl trimethylsilylacetate,methyl 2-trimethylsilyl acetate,acetic acid, trimethylsilyl-, methyl ester,acmc-1ch43,jihuzdffyvrxkp-uhfffaoysa,trimethylsilanyl-acetic acid methyl ester
76210
methyl 2-trimethylsilylacetate
COC(=O)C[Si](C)(C)C
Refractive Index 1.4130 to 1.4150 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5g
Color Colorless
Density 0.8900g/mL
Assay Percent Range 98%
Linear Formula (CH3)3SiCH2CO2CH3
Formula Weight 146.26
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Methyl (trimethylsilyl)acetate, 98%
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning