Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl (trimethylsilyl)propiolate, 98%

Manufacturer:  Alfa Aesar™ B2524514

Quantity 25g

 View more versions of this product

Catalog No. AAB2524514


  Check Availability
Add to cart

Specifications

Description

Applications
Methyl (trimethylsilyl)propiolate is used as the intermediates of OLED, pharmaceuticals.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Description & Specifications

Chemical Identifiers

42201-71-8
(CH3)3SiC≡CCO2CH3
MFCD04039783
Methyl 3-(trimethylsilyl)propynoate; (Trimethylsilyl)propiolic acid methyl ester
C7H12O2Si
156.25
UN3272
Methyl (trimethylsilyl)propiolate

Specifications

42201-71-8
(CH3)3SiC≡CCO2CH3
MFCD04039783
Methyl 3-(trimethylsilyl)propynoate; (Trimethylsilyl)propiolic acid methyl ester
25g
98%
C7H12O2Si
156.25
UN3272
71° to 75°C (11mmHg)
Methyl (trimethylsilyl)propiolate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit