Alfa Aesar™ Methyl vanillate, 99%

Manufacturer: Alfa Aesar™ B2420922

Quantity 100g

 View more versions of this product

Catalog No. AAB2420922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3943-74-6
    Molecular Formula C9H10O4
    Formula Weight 182.18
    MDL Number MFCD00008438
    Beilstein 1369113
    Synonym Methyl 4-hydroxy-3-methoxybenzoate; Vanillic acid methyl ester
    Chemical Name or Material Methyl vanillate

    Specifications

    Formula Weight 182.18
    MDL Number MFCD00008438
    Health Hazard 1 Warning
    Boiling Point 285° to 287°C
    Melting Point 62° to 66°C
    Quantity 100g
    TSCA TSCA
    Molecular Formula C9H10O4
    CAS 3943-74-6
    Beilstein 1369113
    Synonym Methyl 4-hydroxy-3-methoxybenzoate; Vanillic acid methyl ester
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number DH2430000
    Chemical Name or Material Methyl vanillate
    Assay 99%

    Description

    Applications
    Methyl vanillate is used to investigate the volatile compounds of heather honeys from Calluna vulgaris and Erica arborea.

    Solubility
    Soluble in chloroform and methanol.

    Notes
    Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit