Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl vinyl ether/maleic anhydride copolymer, medium molecular weight, Acros Organics

Click to view available options
Quantity:
100g
9011-16-9
C7H8O4
156.137
MFCD00134013
UPBDXRPQPOWRKR-UHFFFAOYSA-N
gantrez,viscofas,pvm/ma copolymer,gantrez an 119,gantrez an,gantrez 39,gantrez 149,gantrez 169,gantrez 903,gantrez s 95
62708
furan-2,5-dione;methoxyethene
COC=C.C1=CC(=O)OC1=O
9011-16-9
C7H8O4
156.137
MFCD00134013
UPBDXRPQPOWRKR-UHFFFAOYSA-N
gantrez,viscofas,pvm/ma copolymer,gantrez an 119,gantrez an,gantrez 39,gantrez 149,gantrez 169,gantrez 903,gantrez s 95
62708
furan-2,5-dione;methoxyethene
COC=C.C1=CC(=O)OC1=O
Additional Information specific viscosity (25°C, c=1, MEK): 1.0 to 1.5
Physical Form Powder
Infrared Spectrum Authentic
Color White
Quantity 100g
Chemical Name or Material Methyl vinyl ether/maleic anhydride copolymer