Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Vinyl Ketone, Stabilized ca 95%, ACROS Organics™

Click to view available options
Quantity:
5mL
2.5L
Packaging:
Glass bottle
78-94-4,75-05-8,64-19-7,123-31-9,7732-18-5
C4H6O
70.091
FUSUHKVFWTUUBE-UHFFFAOYSA-N
methyl vinyl ketone,3-buten-2-one,butenone,methylvinylketon,vinyl methyl ketone,methylene acetone,2-butenone,acetyl ethylene,3-butene-2-one,methyl ethenyl ketone
6570
CHEBI:48058
but-3-en-2-one
CC(=O)C=C
78-94-4,75-05-8,64-19-7,123-31-9,7732-18-5
C4H6O
70.091
FUSUHKVFWTUUBE-UHFFFAOYSA-N
methyl vinyl ketone,3-buten-2-one,butenone,methylvinylketon,vinyl methyl ketone,methylene acetone,2-butenone,acetyl ethylene,3-butene-2-one,methyl ethenyl ketone
6570
CHEBI:48058
but-3-en-2-one
CC(=O)C=C
Density 0.8640g/mL
Infrared Spectrum Authentic
Quantity 5mL
Flash Point −7°C
Stabilizer 0.7% acetonitrile max., 0.4 to 1.0% acetic acid, approx. 0.3% hydroquinone
Refractive Index 1.4100 to 1.4120
Specific Gravity 0.864
Packaging Glass bottle
Water ∽3%
Boiling Point 80.0°C
Color Brown to Colorless
Assay Percent Range ca. 95%
Linear Formula H2C=CHCOCH3
Fieser 01,697; 02,283; 05,464; 06,407; 07,247; 10,272; 12,329; 13,193
Formula Weight 70.09
Physical Form Liquid
Percent Purity ca. 95%
Chemical Name or Material Methyl vinyl ketone
Show More Show Less