Alfa Aesar™ Methyl vinyl ketone, tech. 90%, stab.

Manufacturer: Alfa Aesar™ A11910AP

Quantity 500mL

 View more versions of this product

Catalog No. AAA11910AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 78-94-4
    Molecular Formula C4H6O
    Formula Weight 70.09
    MDL Number MFCD00008777
    UN Number UN1251
    Beilstein 506021
    Merck Index 14,6134
    Synonym 3-Buten-2-one
    Chemical Name or Material Methyl vinyl ketone, Stabilized
    Grade Technical Grade

    Specifications

    Formula Weight 70.09
    Physical Form Liquid
    Assay Percent Range 90%, Stabilized with 0.5% Hydroquinone
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 78-94-4
    Beilstein 506021
    Health Hazard 1 Danger
    Molecular Formula C4H6O
    Boiling Point 32° to 34°C (60mmHg)
    Quantity 500mL
    Recommended Storage 2°C to 8°C
    MDL Number MFCD00008777
    UN Number UN1251
    Synonym 3-Buten-2-one
    Melting Point −7°C
    TSCA TSCA
    Merck Index 14,6134
    Health Hazard 2 GHS H Statement
    H224-H300-H330-H314-H318
    Extremely flammable liquid and vapor.
    Fatal if swallowed.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: METHYL VINYL KETONE, STABILIZED
    RTECS Number EM9800000
    Flash Point −6°C (21°F)
    Chemical Name or Material Methyl vinyl ketone, Stabilized
    Assay 90%
    Grade Technical Grade

    GHS H Statement
    H224-H300-H330-H314-H318
    Extremely flammable liquid and vapor.
    Fatal if swallowed.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    2°C to 8°C

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit