Alfa Aesar™ Methylaminoacetonitrile hydrochloride, 99%

Manufacturer: Alfa Aesar™ H5521514

Quantity 25g

 View more versions of this product

Catalog No. AAH5521514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 25808-30-4
    Molecular Formula C3H6N2·HCl
    Linear Formula CH3NHCH2CN·HCl
    Formula Weight 106.55
    MDL Number MFCD00012587
    Beilstein 3908975
    Synonym Sarcosinenitrile hydrochloride
    Chemical Name or Material Methylaminoacetonitrile hydrochloride

    Specifications

    Formula Weight 106.55
    Linear Formula CH3NHCH2CN·HCl
    Melting Point 103° to 106°C
    Quantity 25g
    TSCA TSCA
    Molecular Formula C3H6N2·HCl
    MDL Number MFCD00012587
    CAS 25808-30-4
    Beilstein 3908975
    Synonym Sarcosinenitrile hydrochloride
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Methylaminoacetonitrile hydrochloride
    Assay 99%

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit