Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methylated Lysine Polyclonal Antibody
Supplier: Thermo Scientific PA5115718
Description

Specifications
Methylated Lysine | |
Polyclonal | |
Unconjugated | |
Dimethylysine; Methyl lysine; Trimethyl lysine | |
A synthesized peptide derived from human Pan Methylated Lysine, corresponding to a region within C-terminal amino acids. | |
100 μL | |
Primary | |
-20°C | |
Liquid | |
Chemical |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Rabbit | |
Affinity Chromatography | |
RUO | |
Chemical | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction