Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methylcobalamin hydrate, 99%

Manufacturer:  Alfa Aesar™ A11176ME

Quantity 500mg

 View more versions of this product

Catalog No. AAA11176ME


  Check Availability
Add to cart

Specifications

Description

Applications
Methylcobalamin hydrate is a coenzyme required in the biosynthesis of methionine. Vitamin (hematopoietic). It acts as a histamine receptor, Alzheimer research.Methylcobalamin is also used in the treatment of peripheral neuropathy, diabetic neuropathy, and as a preliminary treatment for amyotrophic lateral sclerosis. It can be used to prevent or treat pathology arising from a lack of vitamin B12 such as pernicious anemia.

Solubility
Partially soluble in cold water, hot water.

Notes
Store at -20°C. Incompatible with strong oxidizing agents.
Description & Specifications

Chemical Identifiers

13422-55-4
1344.41 (Anhydrous)
Cobinamide; Mecobalamin
C63H91CoN13O14P·xH2O
MFCD00149221
Methylcobalamin hydrate

Specifications

13422-55-4
1344.41 (Anhydrous)
Cobinamide; Mecobalamin
Methylcobalamin hydrate
C63H91CoN13O14P·xH2O
MFCD00149221
500mg
99%

Safety and Handling

SDS & Safety Documents

RTECSNumber : GG3745000

Recommended Storage : −30°C to −10°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit