Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylcyclohexane, 99%, extra pure, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
5L
10L
Packaging:
Glass Bottle
Plastic drum
108-87-2
C7H14
98.19
MFCD00001497
UAEPNZWRGJTJPN-UHFFFAOYSA-N
cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan
7962
methylcyclohexane
CC1CCCCC1
108-87-2
C7H14
98.19
MFCD00001497
UAEPNZWRGJTJPN-UHFFFAOYSA-N
cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan
7962
methylcyclohexane
CC1CCCCC1
Boiling Point 101.0°C
Melting Point -126.3°C
Color Undesignated
Quantity 10L
Infrared Spectrum Authentic
Flash Point −3°C
Packaging Plastic drum
Density 0.7700g/mL
Refractive Index 1.4210 to 1.4230
Specific Gravity 0.77
Assay Percent Range 99%
Linear Formula C6H11CH3
Beilstein 05, 29
Merck Index 15, 6118
Solubility Information Solubility in water: 0.1g/L (20°C). Other solubilities: soluble in acetone,benzene,carbon tetrachloride,,ether,ethanol
Formula Weight 98.19
Physical Form Liquid
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material Methylcyclohexane, 99%
Show More Show Less

GHS H Statement:
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-624-3

RTECSNumber : GV6125000

TSCA : TSCA