Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylcyclopentadiene dimer, 95%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
2.5kg
Packaging:
Glass bottle
26472-00-4
C12H16
160.26
MFCD00082297
26472-00-4
C12H16
160.26
MFCD00082297
Flash Point 26°C
Refractive Index 1.4975 to 1.4995
Quantity 250g
Melting Point -51.0°C
Boiling Point 200.0°C
Density 0.9400g/mL
NMR 94% min.
Color Colorless to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Specific Gravity 0.94
Assay Percent Range 95%
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,benzene and ether
Formula Weight 160.26
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Methylcyclopentadiene dimer
Show More Show Less

GHS H Statement
Flammable liquid and vapour.
Suspected of causing cancer.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 247-724-5