Alfa Aesar™ Methylcyclopentadienylmanganese tricarbonyl, 97%

Manufacturer: Alfa Aesar™ 4712514

Quantity 25g

 View more versions of this product

Catalog No. AA4712514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12108-13-3
    Molecular Formula C9H7MnO3
    Formula Weight 218.09
    MDL Number MFCD00001426
    UN Number UN3281
    Merck Index 14,6223
    Synonym MMT
    Chemical Name or Material Methylcyclopentadienylmanganese tricarbonyl

    Specifications

    Formula Weight 218.09
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Color Yellow
    Health Hazard 1 Danger
    Sensitivity Air Sensitive
    Health Hazard 2 GHS H Statement
    H300-H310-H330
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Density 1.388g/mL
    Refractive Index 1.5873
    MDL Number MFCD00001426
    CAS 12108-13-3
    Synonym MMT
    Solubility Information Insoluble in water; Soluble in hydrocarbon solvents, alcohols, ether, acetone
    Quantity 25g
    Physical Form Liquid
    UN Number UN3281
    Merck Index 14,6223
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, LIQUID, N.O.S.
    RTECS Number OP1450000
    TSCA TSCA
    Molecular Formula C9H7MnO3
    Melting Point 1.5°C
    Boiling Point 233°C
    Flash Point 96°C (205°F)
    Chemical Name or Material Methylcyclopentadienylmanganese tricarbonyl
    Assay 97%

    GHS H Statement
    H300-H310-H330
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit