Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methylcyclopentane, ≥99.5% (GC), MilliporeSigma™ Supelco™
Methylcyclopentane, a naphthene hydrocarbon, has a tertiary carbon atom and also has greater reactivity towards chemical agents like sulphuric acid, fuming nitric acid, etc.
Supplier: MilliporeSigma Supelco 6649050ML
Description
Featured Industries: EnvironmentalSpecifications
96-37-7 | |
Analytical Standard | |
MFCD00001382 | |
≥99.5% (GC) | |
624°F | |
0.748 g/mL (at 20°C) | |
72°C | |
Neat |
50 mL | |
C5H9 CH3 | |
84.16 | |
Analytical Standard | |
232.8 mmHg (37.7°C) | |
-142°C | |
-29.0°C - closed cup |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction