Alfa Aesar™ Methyldiphenylphosphine oxide, 98+%

Manufacturer: Alfa Aesar™ A1148414

Quantity 25g

 View more versions of this product

Catalog No. AAA1148414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2129-89-7
    Molecular Formula C13H13OP
    Formula Weight 216.22
    MDL Number MFCD00002081
    Beilstein 957291
    Synonym Diphenylmethylphosphine oxide
    Chemical Name or Material Methyldiphenylphosphine oxide

    Specifications

    Formula Weight 216.22
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 2129-89-7
    Beilstein 957291
    Synonym Diphenylmethylphosphine oxide
    Quantity 25g
    Molecular Formula C13H13OP
    MDL Number MFCD00002081
    Health Hazard 1 Warning
    Melting Point 111° to 114°C
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number SZ0199480
    Chemical Name or Material Methyldiphenylphosphine oxide
    Assay ≥98%

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit