Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (S)-(-)-lactate, 97%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
2.5kg
Packaging:
Glass bottle
27871-49-4
C4H8O3
104.11
MFCD00064265
LPEKGGXMPWTOCB-VKHMYHEASA-N
methyl s---lactate, methyl l-lactate, s-methyl lactate, --methyl l-lactate, s-lactic acid methyl ester, methyl 2s-2-hydroxypropanoate, --methyl lactate, methyl s-2-hydroxypropionate, propanoic acid, 2-hydroxy-, methyl ester, 2s, --lactic acid methyl ester
94386
CHEBI:83222
methyl (2S)-2-hydroxypropanoate
COC(=O)[C@H](C)O
27871-49-4
C4H8O3
104.11
MFCD00064265
LPEKGGXMPWTOCB-VKHMYHEASA-N
methyl s---lactate, methyl l-lactate, s-methyl lactate, --methyl l-lactate, s-lactic acid methyl ester, methyl 2s-2-hydroxypropanoate, --methyl lactate, methyl s-2-hydroxypropionate, propanoic acid, 2-hydroxy-, methyl ester, 2s, --lactic acid methyl ester
94386
CHEBI:83222
methyl (2S)-2-hydroxypropanoate
COC(=O)[C@H](C)O
Water 0.3% max.
Specific Rotation − 8.50
Specific Gravity 1.092
Specific Rotation Condition − 8.50 (20.00°C neat)
Refractive Index 1.4100 to 1.4150
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 250g
Flash Point 55°C
Color Undesignated
Acidity 0.2% max. (as lactic acid)
Density 1.0920g/mL
Boiling Point 144.0°C to 145.0°C
Viscosity 2.9 mPa.s (25°C)
Assay Percent Range 96% min. (GC)
Linear Formula CH3CH(OH)CO2CH3
Beilstein 03, 264
Solubility Information Solubility in water: miscible,hydrolyses. Other solubilities: miscible with most organic solvents,soluble in alcohol and ether
Formula Weight 104.11
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Methyl (S)-(-)-lactate
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 248-704-9