Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 1-aminocyclopropanecarboxylate hydrochloride, 98%, Acros Organics

Click to view available options
Quantity:
100mg
500mg
72784-42-0
C5H9NO2·ClH
151.59
CHSDCWKLSHPKFY-UHFFFAOYSA-N
methyl 1-aminocyclopropanecarboxylate hydrochloride,1-aminocyclopropane-1-carboxylic acid methyl ester hydrochloride,methyl 1-aminocyclopropane-1-carboxylate hydrochloride,1,1-accp ome,1-amino-cyclopropanecarboxylic acid methyl ester hcl,methyl 1-aminocyclopropane-1-carboxylate hcl,cyclopropanecarboxylic acid, 1-amino-, methyl ester, hydrochloride,ksc498c1l,methyl 1-aminocyclopropanecarboxylate hcl,methyl-1-aminocyclopropanecarboxylate hydrochloride
12760695
methyl 1-aminocyclopropane-1-carboxylate;hydrochloride
COC(=O)C1(CC1)N.Cl
72784-42-0
C5H9NO2·ClH
151.59
CHSDCWKLSHPKFY-UHFFFAOYSA-N
methyl 1-aminocyclopropanecarboxylate hydrochloride,1-aminocyclopropane-1-carboxylic acid methyl ester hydrochloride,methyl 1-aminocyclopropane-1-carboxylate hydrochloride,1,1-accp ome,1-amino-cyclopropanecarboxylic acid methyl ester hcl,methyl 1-aminocyclopropane-1-carboxylate hcl,cyclopropanecarboxylic acid, 1-amino-, methyl ester, hydrochloride,ksc498c1l,methyl 1-aminocyclopropanecarboxylate hcl,methyl-1-aminocyclopropanecarboxylate hydrochloride
12760695
methyl 1-aminocyclopropane-1-carboxylate;hydrochloride
COC(=O)C1(CC1)N.Cl
Melting Point 176.4°C to 178.7°C
Elemental Analysis 38.37 to 40.33% C, 8.95 to 9.40%
Infrared Spectrum Authentic
Color White
Quantity 500mg
Assay Percent Range 98%
Formula Weight 151.59
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Methyl 1-aminocyclopropanecarboxylate hydrochloride

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning