Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,4-dichlorobenzoate, 97%, Acros Organics™

Click to view available options
Quantity:
5g
25g
35112-28-8
C8H6Cl2O2
205.04
MFCD00085117
VCRWILYAWSRHBN-UHFFFAOYSA-N
methyl2,4-dichlorobenzoate,2,4-dichlorobenzoic acid methyl ester,benzoic acid, 2,4-dichloro-, methyl ester,methyl-2,4-dichlorobenzoate,pubchem3713,methyl ester of 2,4-dichlorobenzoic acid,acmc-209id8,ksc497m5b,methyl 2,4-dichloro benzoate
37055
methyl 2,4-dichlorobenzoate
COC(=O)C1=C(C=C(C=C1)Cl)Cl
35112-28-8
C8H6Cl2O2
205.04
MFCD00085117
VCRWILYAWSRHBN-UHFFFAOYSA-N
methyl2,4-dichlorobenzoate,2,4-dichlorobenzoic acid methyl ester,benzoic acid, 2,4-dichloro-, methyl ester,methyl-2,4-dichlorobenzoate,pubchem3713,methyl ester of 2,4-dichlorobenzoic acid,acmc-209id8,ksc497m5b,methyl 2,4-dichloro benzoate
37055
methyl 2,4-dichlorobenzoate
COC(=O)C1=C(C=C(C=C1)Cl)Cl
Color Colorless to Yellow
Boiling Point 132.0°C (15.0 mmHg)
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point 110°C
Quantity 25g
Assay Percent Range 96% min. (GC)
Formula Weight 205.04
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Methyl 2,4-dichlorobenzoate
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 252-374-1