Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-chloroacetoacetate, 95%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
Packaging:
Glass bottle
4755-81-1
C5H7ClO3
150.56
MFCD00008757
GYQRIAVRKLRQKP-UHFFFAOYSA-N
methyl 2-chloroacetoacetate, butanoic acid, 2-chloro-3-oxo-, methyl ester, 2-chloroacetoacetic acid methyl ester, 2-chloro-3-oxo-butyric acid methyl ester, methyl2-chloroacetoacetate, acmc-209k9v, methyl-2-chloroacetoacetate, methyl-alpha-chloroacetoacetate, 2-chloroacetoacetate methyl ester, gyqriavrklrqkp-uhfffaoysa
107332
methyl 2-chloro-3-oxobutanoate
CC(=O)C(C(=O)OC)Cl
4755-81-1
C5H7ClO3
150.56
MFCD00008757
GYQRIAVRKLRQKP-UHFFFAOYSA-N
methyl 2-chloroacetoacetate, butanoic acid, 2-chloro-3-oxo-, methyl ester, 2-chloroacetoacetic acid methyl ester, 2-chloro-3-oxo-butyric acid methyl ester, methyl2-chloroacetoacetate, acmc-209k9v, methyl-2-chloroacetoacetate, methyl-alpha-chloroacetoacetate, 2-chloroacetoacetate methyl ester, gyqriavrklrqkp-uhfffaoysa
107332
methyl 2-chloro-3-oxobutanoate
CC(=O)C(C(=O)OC)Cl
Boiling Point 137°C
Color Yellow
Quantity 10mL
Flash Point 72°C
Melting Point -33°C
Specific Gravity 1.24
Density 1.2400g/mL
Refractive Index 1.4455 to 1.4475
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 95%
Linear Formula CH3COCH(Cl)CO2CH3
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: soluble in organic solvents
Formula Weight 150.56
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Methyl 2-chloroacetoacetate
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 225-286-6