Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-cyclopentanonecarboxylate, 96%, Acros Organics

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
10472-24-9
C7H10O3
142.15
MFCD00001411
PZBBESSUKAHBHD-UHFFFAOYSA-N
methyl 2-oxocyclopentanecarboxylate,methyl 2-cyclopentanonecarboxylate,2-carbmethoxycyclopentanone,methyl cyclopentanone-2-carboxylate,cyclopentanecarboxylic acid, 2-oxo-, methyl ester,2-methoxy carbonyl cyclopentanone,2-methoxycarbonylcyclopentanone,2-oxocyclopentanecarboxylic acid methyl ester,cyclopentanone-2-carboxylic acid methyl ester,mehyl-2-oxocyclopentane
66328
methyl 2-oxocyclopentane-1-carboxylate
COC(=O)C1CCCC1=O
10472-24-9
C7H10O3
142.15
MFCD00001411
PZBBESSUKAHBHD-UHFFFAOYSA-N
methyl 2-oxocyclopentanecarboxylate,methyl 2-cyclopentanonecarboxylate,2-carbmethoxycyclopentanone,methyl cyclopentanone-2-carboxylate,cyclopentanecarboxylic acid, 2-oxo-, methyl ester,2-methoxy carbonyl cyclopentanone,2-methoxycarbonylcyclopentanone,2-oxocyclopentanecarboxylic acid methyl ester,cyclopentanone-2-carboxylic acid methyl ester,mehyl-2-oxocyclopentane
66328
methyl 2-oxocyclopentane-1-carboxylate
COC(=O)C1CCCC1=O
Refractive Index 1.4550 to 1.457
Density 1.0540g/mL
Flash Point 77°C
Boiling Point 102°C to 104°C (11.0mmHg)
Specific Gravity 1.054
Quantity 5g
Color Colorless to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 95% min. (GC)
Beilstein 10, 597
Solubility Information Solubility in water: >100g/L. Other solubilities: miscible in ethanol
Formula Weight 142.15
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material Methyl 2-cyclopentanonecarboxylate
Show More Show Less

EINECSNumber : 233-962-7