Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-amino-2-thiophenecarboxylate, 99%, Acros Organics

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
22288-78-4
C6H7NO2S
157.19
MFCD00009765
TWEQNZZOOFKOER-UHFFFAOYSA-N
methyl 3-amino-2-thiophenecarboxylate,3-aminothiophene-2-carboxylic acid methyl ester,2-thiophenecarboxylic acid, 3-amino-, methyl ester,3-amino-thiophene-2-carboxylic acid methyl ester,3-amino-2-thiophenecarboxylic acid methyl ester,methyl 3-amino-2-thiophene carboxylate,methyl-3-amino-thiophene-2-carboxylate,3-amino-2-carbomethoxythiophene,3-amino-2-methoxycarbonylthiophene,methyl3-aminothiophene-2-carboxylate
89652
methyl 3-aminothiophene-2-carboxylate
COC(=O)C1=C(C=CS1)N
22288-78-4
C6H7NO2S
157.19
MFCD00009765
TWEQNZZOOFKOER-UHFFFAOYSA-N
methyl 3-amino-2-thiophenecarboxylate,3-aminothiophene-2-carboxylic acid methyl ester,2-thiophenecarboxylic acid, 3-amino-, methyl ester,3-amino-thiophene-2-carboxylic acid methyl ester,3-amino-2-thiophenecarboxylic acid methyl ester,methyl 3-amino-2-thiophene carboxylate,methyl-3-amino-thiophene-2-carboxylate,3-amino-2-carbomethoxythiophene,3-amino-2-methoxycarbonylthiophene,methyl3-aminothiophene-2-carboxylate
89652
methyl 3-aminothiophene-2-carboxylate
COC(=O)C1=C(C=CS1)N
Melting Point 66°C to 67°C
Boiling Point 100°C to 102°C (0.1mmHg)
Quantity 10g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Beige to Beige-Brown
Assay Percent Range 99%
Formula Weight 157.19
Physical Form Fine Crystalline Powder
Percent Purity 99%
Chemical Name or Material Methyl 3-amino-2-thiophenecarboxylate
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 244-895-8