Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-amino-5,6-dichloro-2-pyrazinecarboxylate, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
1458-18-0
C6H5Cl2N3O2
222.025
MFCD00010431
USYMCUGEGUFUBI-UHFFFAOYSA-N
methyl 3-amino-5,6-dichloro-2-pyrazinecarboxylate,pyrazinecarboxylic acid, 3-amino-5,6-dichloro-, methyl ester,3-amino-5,6-dichloro-2-pyrazinecarboxylic acid methyl ester,3-amino-5,6-dichloro-pyrazine-2-carboxylic acid methyl ester,acmc-1bujh,dsstox_cid_31434,dsstox_rid_97320,dsstox_gsid_57645,methyl 3-amino-5,6-dichloropyrazinoate,methyl 3-amino-5,6-dichloropyrazinecarboxylate
73828
methyl 3-amino-5,6-dichloropyrazine-2-carboxylate
COC(=O)C1=C(N=C(C(=N1)Cl)Cl)N
1458-18-0
C6H5Cl2N3O2
222.025
MFCD00010431
USYMCUGEGUFUBI-UHFFFAOYSA-N
methyl 3-amino-5,6-dichloro-2-pyrazinecarboxylate,pyrazinecarboxylic acid, 3-amino-5,6-dichloro-, methyl ester,3-amino-5,6-dichloro-2-pyrazinecarboxylic acid methyl ester,3-amino-5,6-dichloro-pyrazine-2-carboxylic acid methyl ester,acmc-1bujh,dsstox_cid_31434,dsstox_rid_97320,dsstox_gsid_57645,methyl 3-amino-5,6-dichloropyrazinoate,methyl 3-amino-5,6-dichloropyrazinecarboxylate
73828
methyl 3-amino-5,6-dichloropyrazine-2-carboxylate
COC(=O)C1=C(N=C(C(=N1)Cl)Cl)N
Quantity 1g
Melting Point 229°C to 235°C (decomposition)
Packaging Glass bottle
Color Brown to Brown-Red
Infrared Spectrum Authentic
Assay Percent Range 98%
Formula Weight 222.03
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material Methyl 3-amino-5, 6-dichloro-2-pyrazinecarboxylate, 98%

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning