Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-aminobenzo[b]thiophene-2-carboxylate, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
35212-85-2
C10H9NO2S
207.25
MFCD00206744
VLHHEYMZLXKSQO-UHFFFAOYSA-N
methyl 3-aminobenzo b thiophene-2-carboxylate,benzo b thiophene-2-carboxylic acid, 3-amino-, methyl ester,cbmicro_030710,2-carbomethoxy-3-aminobenzothiophene,methyl 3-aminobenzothiophene-2-carboxylate,methyl-3-aminobenzothiophene-2-carboxylate,3-amino-2-methoxycarbonyl-1-benzothiophene,methyl 3-amino-benzo b thiophene-2-carboxylate,methyl 3-azanyl-1-benzothiophene-2-carboxylate,amino-benzo b thiophene-2-carboxylic acid methyl ester
874720
methyl 3-amino-1-benzothiophene-2-carboxylate
COC(=O)C1=C(C2=CC=CC=C2S1)N
35212-85-2
C10H9NO2S
207.25
MFCD00206744
VLHHEYMZLXKSQO-UHFFFAOYSA-N
methyl 3-aminobenzo b thiophene-2-carboxylate,benzo b thiophene-2-carboxylic acid, 3-amino-, methyl ester,cbmicro_030710,2-carbomethoxy-3-aminobenzothiophene,methyl 3-aminobenzothiophene-2-carboxylate,methyl-3-aminobenzothiophene-2-carboxylate,3-amino-2-methoxycarbonyl-1-benzothiophene,methyl 3-amino-benzo b thiophene-2-carboxylate,methyl 3-azanyl-1-benzothiophene-2-carboxylate,amino-benzo b thiophene-2-carboxylic acid methyl ester
874720
methyl 3-amino-1-benzothiophene-2-carboxylate
COC(=O)C1=C(C2=CC=CC=C2S1)N
Melting Point 109°C to 111°C
Color Yellow to Brown
Packaging Glass bottle
Quantity 1g
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Formula Weight 207.25
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material Methyl 3-aminobenzo[b]thiophene-2-carboxylate

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning