Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-(bromomethyl)benzoate, 95%, Acros Organics

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
1129-28-8
C9H9BrO2
229
MFCD00051437
YUHSMQQNPRLEEJ-UHFFFAOYSA-N
methyl 3-bromomethyl benzoate,m-carbomethoxybenzyl bromide,3-bromomethyl benzoic acid methyl ester,benzoic acid, 3-bromomethyl-, methyl ester,methyl-3-bromomethylbenzoate,methyl alpha-bromo-m-toluate,methyl3-bromomethyl benzoate,methyl-3-bromomethyl benzoate,3-carbomethoxybenzyl bromide
517981
methyl 3-(bromomethyl)benzoate
COC(=O)C1=CC(=CC=C1)CBr
1129-28-8
C9H9BrO2
229
MFCD00051437
YUHSMQQNPRLEEJ-UHFFFAOYSA-N
methyl 3-bromomethyl benzoate,m-carbomethoxybenzyl bromide,3-bromomethyl benzoic acid methyl ester,benzoic acid, 3-bromomethyl-, methyl ester,methyl-3-bromomethylbenzoate,methyl alpha-bromo-m-toluate,methyl3-bromomethyl benzoate,methyl-3-bromomethyl benzoate,3-carbomethoxybenzyl bromide
517981
methyl 3-(bromomethyl)benzoate
COC(=O)C1=CC(=CC=C1)CBr
Quantity 5g
Color White
Melting Point 35.0°C to 37.0°C
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point >110°C
Boiling Point 135.0°C (2.0 mmHg)
Assay Percent Range 94% min. (GC)
Linear Formula CH2BrC6H4C(O)OCH3
Beilstein 09, IV, 1721
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 229
Physical Form Crystals or Crystalline Powder
Percent Purity 95%
Chemical Name or Material Methyl 3-(bromomethyl)benzoate
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger