Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-chlorobenzoate, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
2905-65-9
C8H7ClO2
170.592
MFCD00000592
XRDRKVPNHIWTBX-UHFFFAOYSA-N
methyl m-chlorobenzoate,benzoic acid, 3-chloro-, methyl ester,m-chlorobenzoic acid methyl ester,methyl-3-chlorobenzoate,benzoic acid, m-chloro-, methyl ester,3-chlorobenzoic acid methyl ester,3-chloro-benzoic acid methyl ester,acmc-1cegg,dsstox_cid_7507,3-chlorobenzoic acid methyl
17946
methyl 3-chlorobenzoate
COC(=O)C1=CC(=CC=C1)Cl
2905-65-9
C8H7ClO2
170.592
MFCD00000592
XRDRKVPNHIWTBX-UHFFFAOYSA-N
methyl m-chlorobenzoate,benzoic acid, 3-chloro-, methyl ester,m-chlorobenzoic acid methyl ester,methyl-3-chlorobenzoate,benzoic acid, m-chloro-, methyl ester,3-chlorobenzoic acid methyl ester,3-chloro-benzoic acid methyl ester,acmc-1cegg,dsstox_cid_7507,3-chlorobenzoic acid methyl
17946
methyl 3-chlorobenzoate
COC(=O)C1=CC(=CC=C1)Cl
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless to Yellow
Quantity 100g
Refractive Index 1.5310 to 1.5340 (20°C, 589nm)
Density 104g/mL
Assay Percent Range 98%
Linear Formula ClC6H4CO2CH3
Beilstein 09, 338
Merck Index 10, 2093
Formula Weight 170.6
Physical Form Liquid or Low Melting Solid
Percent Purity ≥97.5%
Chemical Name or Material Methyl 3-chlorobenzoate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.