Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-formylbenzoate, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
52178-50-4
C9H8O3
164.2
MFCD00189379
UVSBCUAQEZINCQ-UHFFFAOYSA-N
3-carbomethoxybenzaldehyde,methyl m-formylbenzoate,benzoic acid, 3-formyl-, methyl ester,3-formylbenzoic acid methyl ester,methyl-3-formylbenzoate,pubchem12631,m-carbomethoxybenzaldehyde,methyl 3-formyl-benzoate,methyl 3-methanoylbenzoate,acmc-1am8j
639145
methyl 3-formylbenzoate
COC(=O)C1=CC=CC(=C1)C=O
52178-50-4
C9H8O3
164.2
MFCD00189379
UVSBCUAQEZINCQ-UHFFFAOYSA-N
3-carbomethoxybenzaldehyde,methyl m-formylbenzoate,benzoic acid, 3-formyl-, methyl ester,3-formylbenzoic acid methyl ester,methyl-3-formylbenzoate,pubchem12631,m-carbomethoxybenzaldehyde,methyl 3-formyl-benzoate,methyl 3-methanoylbenzoate,acmc-1am8j
639145
methyl 3-formylbenzoate
COC(=O)C1=CC=CC(=C1)C=O
Melting Point 48.0°C to 55.0°C
Quantity 1g
Packaging Glass bottle
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material Methyl 3-formylbenzoate, 98%

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning