Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl 4-(bromomethyl)benzoate, 98%, ACROS Organics™
Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
Specifications
Infrared Spectrum | Authentic |
Boiling Point | 130.0°C to 135.0°C (2.0mmHg) |
Melting Point | 53.0°C to 58.0°C |
Packaging | Glass bottle |
Quantity | 5g |
Color | White to Yellow |
Assay Percent Range | 97.5% min. (GC) |
Linear Formula | BrCH2C6H4CO2CH3 |
Formula Weight | 229.07 |
Physical Form | Crystalline Powder, Crystals and/or Chunks |
Show More |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger