Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-cyanobenzoate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
1129-35-7
C9H7NO2
161.16
KKZMIDYKRKGJHG-UHFFFAOYSA-N
4-cyanobenzoic acid methyl ester,benzoic acid, 4-cyano-, methyl ester,methyl p-cyanobenzoate,unii-f8fms7o8xg,f8fms7o8xg,benzoic acid, p-cyano-, methyl ester,4-cyano-benzoic acid methyl ester,methyl4-cyanobenzoate,methyl-4-cyanobenzoate,acmc-1bs5i
70791
methyl 4-cyanobenzoate
COC(=O)C1=CC=C(C=C1)C#N
1129-35-7
C9H7NO2
161.16
KKZMIDYKRKGJHG-UHFFFAOYSA-N
4-cyanobenzoic acid methyl ester,benzoic acid, 4-cyano-, methyl ester,methyl p-cyanobenzoate,unii-f8fms7o8xg,f8fms7o8xg,benzoic acid, p-cyano-, methyl ester,4-cyano-benzoic acid methyl ester,methyl4-cyanobenzoate,methyl-4-cyanobenzoate,acmc-1bs5i
70791
methyl 4-cyanobenzoate
COC(=O)C1=CC=C(C=C1)C#N
Melting Point 67.0°C to 68.0°C
Boiling Point 142.0°C to 144.0°C (12.0mmHg)
Infrared Spectrum Authentic
Quantity 5g
Color White
Assay Percent Range 97.5% min. (GC)
Linear Formula NCC6H4CO2CH3
Beilstein 09,846
Formula Weight 161.16
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Methyl 4-cyanobenzoate
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 214-443-4