Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-methoxysalicylate, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
5446-02-6
C9H10O4
182.18
MFCD00008424
ZICRWXFGZCVTBZ-UHFFFAOYSA-N
Methyle2-hydroxy-4-methoxybenzoate
methyl 2-hydroxy-4-methoxybenzoate
COC(=O)C1=CC=C(OC)C=C1O
5446-02-6
C9H10O4
182.18
MFCD00008424
ZICRWXFGZCVTBZ-UHFFFAOYSA-N
Methyle2-hydroxy-4-methoxybenzoate
methyl 2-hydroxy-4-methoxybenzoate
COC(=O)C1=CC=C(OC)C=C1O
Quantity 50g
Flash Point >112°C
Melting Point 49°C to 53°C
Color White to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3OC6H3(OH)CO2CH3
Beilstein 10, 381
Formula Weight 182.18
Physical Form Crystalline Powder or Crystals
Percent Purity 98%
Chemical Name or Material Methyl 4-methoxysalicylate
Show More Show Less

EINECSNumber : 226-657-5