Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyle5-bromonicotinate, 98%, Acros Organics

Click to view available options
Quantity:
1g
29681-44-5
C7H6BrNO2
216.034
MFCD00173668
AAJZXPWBILCHAW-UHFFFAOYSA-N
methyl 5-bromonicotinate,5-bromonicotinic acid methyl ester,5-bromo-3-pyridinecarboxylic acid methyl ester,5-bromo nicotinic acid methyl ester,3-pyridinecarboxylic acid, 5-bromo-, methyl ester,3-bromo-5-methoxycarbonyl pyridine,5-bromopyridine-3-carboxylic acid methyl ester,5-bromo-nicotinic acid methyl ester,methyl 5-bromo-3-pyridinecarboxylate,methyl 3-bromo-5-pyridinecarboxylate
699336
methyl 5-bromopyridine-3-carboxylate
COC(=O)C1=CC(=CN=C1)Br
29681-44-5
C7H6BrNO2
216.034
MFCD00173668
AAJZXPWBILCHAW-UHFFFAOYSA-N
methyl 5-bromonicotinate,5-bromonicotinic acid methyl ester,5-bromo-3-pyridinecarboxylic acid methyl ester,5-bromo nicotinic acid methyl ester,3-pyridinecarboxylic acid, 5-bromo-, methyl ester,3-bromo-5-methoxycarbonyl pyridine,5-bromopyridine-3-carboxylic acid methyl ester,5-bromo-nicotinic acid methyl ester,methyl 5-bromo-3-pyridinecarboxylate,methyl 3-bromo-5-pyridinecarboxylate
699336
methyl 5-bromopyridine-3-carboxylate
COC(=O)C1=CC(=CN=C1)Br
Melting Point 97°C to 101°C
Color White
Quantity 1g
Infrared Spectrum Authentic
Assay Percent Range 98%
Beilstein 22, V,2, 181
Formula Weight 216.03
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material Methyle5-bromonicotinate, 98%

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.