Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 6-(trifluoromethyl)nicotinate, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
221313-10-6
C8H6F3NO2
205.13
MFCD03001346
NEATXKNYTRXHQL-UHFFFAOYSA-N
methyl 6-trifluoromethyl nicotinate,methyl 6-trifluoromethyl pyridine-3-carboxylate,methyl6-trifluoromethyl nicotinate,3-pyridinecarboxylic acid, 6-trifluoromethyl-, methyl ester,methyl 6-trifluoromethylnicotinate,methyl 2-trifluoromethyl pyridine-5-carboxylate,methyl 2-trifluoromethyl-5-pyridinecarboxylate,6-trifluoromethylnicotinic acid methyl ester,pubchem18762,methyltrifluoromethylnicotinate
2775647
methyl 6-(trifluoromethyl)pyridine-3-carboxylate
COC(=O)C1=CN=C(C=C1)C(F)(F)F
221313-10-6
C8H6F3NO2
205.13
MFCD03001346
NEATXKNYTRXHQL-UHFFFAOYSA-N
methyl 6-trifluoromethyl nicotinate,methyl 6-trifluoromethyl pyridine-3-carboxylate,methyl6-trifluoromethyl nicotinate,3-pyridinecarboxylic acid, 6-trifluoromethyl-, methyl ester,methyl 6-trifluoromethylnicotinate,methyl 2-trifluoromethyl pyridine-5-carboxylate,methyl 2-trifluoromethyl-5-pyridinecarboxylate,6-trifluoromethylnicotinic acid methyl ester,pubchem18762,methyltrifluoromethylnicotinate
2775647
methyl 6-(trifluoromethyl)pyridine-3-carboxylate
COC(=O)C1=CN=C(C=C1)C(F)(F)F
Melting Point 58°C to 60.5°C
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Assay Percent Range 96% min. (GC)
Formula Weight 205.13
Physical Form Powder
Percent Purity 97%
Chemical Name or Material Methyl 6-(trifluoromethyl)nicotinate

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning