Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl benzenesulfinate, 98%, Acros Organics™

Click to view available options
Quantity:
5mL
25mL
670-98-4
C7H8O2S
156.2
PSNSVDSRLUYDKF-UHFFFAOYSA-N
methylbenzenesulfinate,benzenesulfinic acid methyl ester,methyl phenylsulfinate,methyl benzene sulfinate,benzenesulfinic acid,methyl ester,benzenesulfinic acid, methyl ester, r,benzenesulfinic acid, methyl ester, s,methoxy phenylsulfinyl,acmc-20mem2,acmc-20mg8f
2759784
methyl benzenesulfinate
COS(=O)C1=CC=CC=C1
670-98-4
C7H8O2S
156.2
PSNSVDSRLUYDKF-UHFFFAOYSA-N
methylbenzenesulfinate,benzenesulfinic acid methyl ester,methyl phenylsulfinate,methyl benzene sulfinate,benzenesulfinic acid,methyl ester,benzenesulfinic acid, methyl ester, r,benzenesulfinic acid, methyl ester, s,methoxy phenylsulfinyl,acmc-20mem2,acmc-20mg8f
2759784
methyl benzenesulfinate
COS(=O)C1=CC=CC=C1
Boiling Point 110.0°C to 112.0°C (16.0 Torr)
Color Colorless to Yellow
Density 1.1940g/mL
Specific Gravity 1.194
Infrared Spectrum Authentic
Quantity 25mL
Assay Percent Range 97.5% min. (GC)
Formula Weight 156.2
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Methyl benzenesulfinate
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call

GHS Signal Word: Warning