Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl cyclopropanecarboxylate, 98%, Acros Organics™

Click to view available options
Quantity:
25g
100g
500g
2868-37-3
C5H8O2
100.12
MFCD00001280
PKAHQJNJPDVTDP-UHFFFAOYSA-N
methyl cyclopropane carboxylate,cyclopropanecarboxylic acid, methyl ester,methylcyclopropanecarboxylate,cyclopropanecarboxylic acid methyl ester,methyl cyclopropylcarboxylate,unii-5uq53f585u,mcpc,pubchem11835,ksc490a7l,1-methylcyclopropanecarboxylate
76122
methyl cyclopropanecarboxylate
COC(=O)C1CC1
2868-37-3
C5H8O2
100.12
MFCD00001280
PKAHQJNJPDVTDP-UHFFFAOYSA-N
methyl cyclopropane carboxylate,cyclopropanecarboxylic acid, methyl ester,methylcyclopropanecarboxylate,cyclopropanecarboxylic acid methyl ester,methyl cyclopropylcarboxylate,unii-5uq53f585u,mcpc,pubchem11835,ksc490a7l,1-methylcyclopropanecarboxylate
76122
methyl cyclopropanecarboxylate
COC(=O)C1CC1
Color Colorless
Infrared Spectrum Authentic
Quantity 500g
Flash Point 17°C
Density 0.9800g/mL
Refractive Index 1.4170 to 1.4190
Specific Gravity 0.98
Packaging Glass bottle
Boiling Point 119.0°C (764.0 mmHg)
Assay Percent Range 98%
Linear Formula C3H5CO2CH3
Beilstein 09, I, 3
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohols
Formula Weight 100.12
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Methyl cyclopropanecarboxylate
Show More Show Less

GHS P Statement
Causes serious eye irritation.
Toxic in contact with skin.
Harmful if swallowed.
Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger