Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl isothiocyanate, 97%, AcroSeal™, Acros Organics™

Click to view available options
Quantity:
100mL
Packaging:
AcroSeal™ Glass bottle
556-61-6
C2H3NS
73.113
MFCD00004818
LGDSHSYDSCRFAB-UHFFFAOYSA-N
methyl isothiocyanate,isothiocyanatomethane,methane, isothiocyanato,trapex,methyl mustard oil,methyl mustard,mitc,mencs,methylisothiocyanate,trapexide
11167
CHEBI:78337
methylimino(sulfanylidene)methane
CN=C=S
556-61-6
C2H3NS
73.113
MFCD00004818
LGDSHSYDSCRFAB-UHFFFAOYSA-N
methyl isothiocyanate,isothiocyanatomethane,methane, isothiocyanato,trapex,methyl mustard oil,methyl mustard,mitc,mencs,methylisothiocyanate,trapexide
11167
CHEBI:78337
methylimino(sulfanylidene)methane
CN=C=S
Infrared Spectrum Authentic
Color Orange-Yellow to Yellow
Boiling Point 117.0°C to 120.0°C
Packaging AcroSeal™ Glass bottle
Quantity 100mL
Flash Point 32°C
Density 1.0690g/mL
Melting Point 30.0°C to 36.0°C
Specific Gravity 1.069
Assay Percent Range 97%
Linear Formula CH3NCS
Formula Weight 73.12
Physical Form Low Melting Solid
Percent Purity 97%
Chemical Name or Material Methyl isothiocyanate
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Toxic if inhaled.
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger