Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylenaminoacetonitrile, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
109-82-0
C3H4N2
68.079
MFCD00001889
GFZMFCVDDFHSJK-UHFFFAOYSA-N
methyleneaminoacetonitrile,methylenaminoacetonitrile,maan,acetonitrile, methyleneamino,methyleniminoacetonitrile,glycinonitrile, n-methylene,n-methyleneglycinonitrile,2-methylideneamino acetonitrile,methylenamino acetonitrile,usaf do-5
8015
2-(methylideneamino)acetonitrile
C=NCC#N
109-82-0
C3H4N2
68.079
MFCD00001889
GFZMFCVDDFHSJK-UHFFFAOYSA-N
methyleneaminoacetonitrile,methylenaminoacetonitrile,maan,acetonitrile, methyleneamino,methyleniminoacetonitrile,glycinonitrile, n-methylene,n-methyleneglycinonitrile,2-methylideneamino acetonitrile,methylenamino acetonitrile,usaf do-5
8015
2-(methylideneamino)acetonitrile
C=NCC#N
Quantity 5g
Infrared Spectrum Authentic
Melting Point 127°C to 130°C
Color White
Assay Percent Range 98%
Merck Index 15, 6129
Formula Weight 68.07
Physical Form Crystals
Percent Purity ≥97.5%
Chemical Name or Material Methylenaminoacetonitrile, 98%

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning