Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methylene Blue, 1% (w/v) in Denatured Alcohol, Ricca Chemical
Manufacturer: Ricca Chemical Company 486432
Specifications
Methylene Blue | |
97.0, 1.21 | |
7220-79-3, 67-56-1 | |
Poly natural | |
Blue |
1% (w/v) | |
100.0, 1.26 | |
Liquid | |
Denatured Alcohol | |
1L |
Promotions
-
Ricca Chemical Company
,
Fisher Scientific
,
Fisherbrand
Spend $250, Get $250 on Ricca Chemical Products
Buy $250 of Ricca Chemical Company products and get $250 more to spend on Ricca items at no additional cost.: 06-30-2021
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title